3-Brom-4'-chlorbenzophenon
Lieferant: FLUOROCHEM
Synonyme:
(3-Bromophenyl)(4-chlorophenyl)methanone
022811-1G
FLUO022811-1GEA
740
EUR
FLUO022811-1G
3-Brom-4'-chlorbenzophenon
3-Brom-4'-chlorbenzophenon
Formel:
C₁₃H₈BrClO Molecular Weight: 295,559 g/mol |
MDL:
MFCD06201572 CAS-Nummer: 75762-56-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...