3-Brom-3'-chlorbenzophenon

Lieferant: FLUOROCHEM

022812-1G
FLUO022812-1GEA 830 EUR
FLUO022812-1G
3-Brom-3'-chlorbenzophenon
3-Brom-3'-chlorbenzophenon
Formel: C₁₃H₈BrClO
MDL: MFCD06201573
CAS-Nummer: 75762-59-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR