3-Brombenzophenon
Lieferant: FLUOROCHEM
Synonyme:
m-Brombenzophenon
022813-1G
FLUO022813-1GEA
82.1
EUR
FLUO022813-1G
3-Brombenzophenon
3-Brombenzophenon
Formel:
C₁₃H₉BrO Molecular Weight: 261,12 g/mol Siedepunkt: 355 °C (1013 hPa) Schmelzpunkt: 74,5…77.5 °C Dichte: 1,42 g/cm³ (20 °C) |
MDL:
MFCD00672015 CAS-Nummer: 1016-77-9 EINECS: 213-808-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...