4,4'-Di-tert-butylbenzophenon

Lieferant: FLUOROCHEM

Synonyme: Bis(4-(tert-butyl)phenyl)methanone

022814-1G
FLUO022814-1GEA 183 EUR
FLUO022814-1G
4,4'-Di-tert-butylbenzophenon
4,4'-Di-tert-butylbenzophenon
Formel: C₂₁H₂₆O
Molecular Weight: 294,4306 g/mol
MDL: MFCD01319937
CAS-Nummer: 15796-82-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR