2-Fluor-4'-methoxybenzophenon

Lieferant: FLUOROCHEM

022815-1G
FLUO022815-1GEA 627 EUR
FLUO022815-1G
2-Fluor-4'-methoxybenzophenon
2-Fluor-4'-methoxybenzophenon
Formel: C₁₄H₁₁FO₂
MDL: MFCD00749331
CAS-Nummer: 66938-29-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR