3-Chlor-3',4',5'-trifluorbenzophenon

Lieferant: FLUOROCHEM

022816-1G
FLUO022816-1GEA 647 EUR
FLUO022816-1G
3-Chlor-3',4',5'-trifluorbenzophenon
3-Chlor-3',4',5'-trifluorbenzophenon
Formel: C₁₃H₆ClF₃O
MDL: MFCD01319931
CAS-Nummer: 746651-95-6
EINECS: 217-300-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR