4-Brom-4'-fluorbenzophenon
Lieferant: FLUOROCHEM
Synonyme:
(4-Bromophenyl)(4-fluorophenyl)methanone
022818-1G
FLUO022818-1GEA
134
EUR
FLUO022818-1G
4-Brom-4'-fluorbenzophenon
4-Brom-4'-fluorbenzophenon
Formel:
C₁₃H₈BrFO Molecular Weight: 279,1044 g/mol |
MDL:
MFCD00672035 CAS-Nummer: 2069-41-2 EINECS: 218-193-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...