4-Chlor-3',4',5'-trifluorbenzophenon
Lieferant: FLUOROCHEM
022820-1G
FLUO022820-1GEA
647
EUR
FLUO022820-1G
4-Chlor-3',4',5'-trifluorbenzophenon
4-Chlor-3',4',5'-trifluorbenzophenon
Formel:
C₁₃H₆ClF₃O |
MDL:
MFCD01319932 CAS-Nummer: 746651-96-7 EINECS: 217-300-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...