4-Butyl-4'-tert-butylbenzophenon
Lieferant: FLUOROCHEM
022821-1G
FLUO022821-1GEA
647
EUR
FLUO022821-1G
4-Butyl-4'-tert-butylbenzophenon
4-Butyl-4'-tert-butylbenzophenon
Formel:
C₂₁H₂₆O |
MDL:
MFCD01319939 CAS-Nummer: 95282-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...