3-Chlor-3',5'-difluorbenzophenon

Lieferant: FLUOROCHEM

022822-1G
FLUO022822-1GEA 647 EUR
FLUO022822-1G
3-Chlor-3',5'-difluorbenzophenon
3-Chlor-3',5'-difluorbenzophenon
Formel: C₁₃H₇ClF₂O
MDL: MFCD01319934
CAS-Nummer: 746651-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR