3,3'-Dichlorbenzophenon

Lieferant: FLUOROCHEM

Synonyme: 3,3-DBP, Bis(3-chlorphenyl)keton

022824-1G
FLUO022824-1GEA 290 EUR
FLUO022824-1G
3,3'-Dichlorbenzophenon
3,3'-Dichlorbenzophenon
Formel: C₁₃H₈Cl₂O
Molecular Weight: 251,11 g/mol
MDL: MFCD00045192
CAS-Nummer: 7094-34-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR