Benzyl-3-chlorphenylketon
Lieferant: FLUOROCHEM
022847-1G
FLUO022847-1GEA
594
EUR
FLUO022847-1G
Benzyl-3-chlorphenylketon
Benzyl-3-chlorphenylketon
Formel:
C₁₄H₁₁ClO |
MDL:
MFCD02260665 CAS-Nummer: 62482-45-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...