Benzyl-4-chlorphenylketon
Lieferant: FLUOROCHEM
Synonyme:
Benzyl 4-chlorophenyl ketone, 4'-Chloro-2-phenylacetophenone
022848-1G
FLUO022848-1GEA
29.3
EUR
FLUO022848-1G
Benzyl-4-chlorphenylketon
Benzyl-4-chlorphenylketon
Formel:
C₁₄H₁₁ClO Molecular Weight: 230,69 g/mol |
MDL:
MFCD00016342 CAS-Nummer: 1889-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...