3',4'-Ethylendioxy-2-phenylacetophenon

Lieferant: FLUOROCHEM

022851-1G
FLUO022851-1GEA 0 EUR
FLUO022851-1G
3',4'-Ethylendioxy-2-phenylacetophenon
3',4'-Ethylendioxy-2-phenylacetophenon
Formel: C₁₆H₁₄O₃
MDL: MFCD01333467
CAS-Nummer: 18212-83-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR