2-(3-Methoxyphenyl)acetophenon

Lieferant: FLUOROCHEM

022857-1G
FLUO022857-1GEA 594 EUR
FLUO022857-1G
2-(3-Methoxyphenyl)acetophenon
2-(3-Methoxyphenyl)acetophenon
Formel: C₁₅H₁₄O₂
MDL: MFCD00446972
CAS-Nummer: 29955-26-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR