2-(4-Methoxyphenyl)acetophenon
Lieferant: FLUOROCHEM
022858-1G
FLUO022858-1GEA 558 EUR
FLUO022858-1G
2-(4-Methoxyphenyl)acetophenon
2-(4-Methoxyphenyl)acetophenon
Formel: C₁₅H₁₄O₂
Molecular Weight: 226,27 g/mol
MDL: MFCD00096294
CAS-Nummer: 24845-40-7