Benzyl-3-methoxyphenylketon

Lieferant: FLUOROCHEM

022859-1G
FLUO022859-1GEA 594 EUR
FLUO022859-1G
Benzyl-3-methoxyphenylketon
Benzyl-3-methoxyphenylketon
Formel: C₁₅H₁₄O₂
MDL: MFCD02260672
CAS-Nummer: 62381-24-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR