(2-Bromphenyl)aceton

Lieferant: FLUOROCHEM

Synonyme: 1-(2-Bromophenyl)propan-2-one

022866-1G
FLUO022866-1GEA 31.1 EUR
FLUO022866-1G
(2-Bromphenyl)aceton
(2-Bromphenyl)aceton
Formel: C₉H₉BrO
Molecular Weight: 213,07 g/mol
MDL: MFCD03410431
CAS-Nummer: 21906-31-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR