(4-Bromphenyl)aceton

Lieferant: FLUOROCHEM

Synonyme: 4-Bromophenylacetone

022868-1G
FLUO022868-1GEA 30.6 EUR
FLUO022868-1G
(4-Bromphenyl)aceton
(4-Bromphenyl)aceton
Formel: C₉H₉BrO
Molecular Weight: 213,07 g/mol
MDL: MFCD00210401
CAS-Nummer: 6186-22-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR