(3-Chlorphenyl)aceton

Lieferant: FLUOROCHEM

Synonyme: 1-(3-Chlorophenyl)propan-2-one

022869-1G
FLUO022869-1GEA 0 EUR
FLUO022869-1G
(3-Chlorphenyl)aceton
(3-Chlorphenyl)aceton
Formel: C₉H₉ClO
Molecular Weight: 168,62 g/mol
MDL: MFCD00082872
CAS-Nummer: 14123-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR