(2-Fluorphenyl)aceton
Lieferant: FLUOROCHEM
Synonyme:
2-Fluorophenylacetone, 1-(2-Fluorophenyl)propan-2-one, 1-(2-Fluorophenyl)-2-propanone
022873-1G
FLUO022873-1GEA
0
EUR
FLUO022873-1G
(2-Fluorphenyl)aceton
(2-Fluorphenyl)aceton
Formel:
C₉H₉FO Siedepunkt: 47 °C (1013 hPa) Dichte: 1,07 g/cm³ (20 °C) |
MDL:
MFCD00000325 CAS-Nummer: 2836-82-0 EINECS: 220-627-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...