(3-Fluorphenyl)aceton

Lieferant: FLUOROCHEM

Synonyme: 1-(3-Fluorophenyl)propan-2-one

022874-1G
FLUO022874-1GEA 93.6 EUR
FLUO022874-1G
(3-Fluorphenyl)aceton
(3-Fluorphenyl)aceton
Formel: C₉H₉FO
Molecular Weight: 152,17 g/mol
MDL: MFCD00061144
CAS-Nummer: 1737-19-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR