2-Phenyl-4'-(trifluormethyl)acetophenon

Lieferant: FLUOROCHEM

022876-1G
FLUO022876-1GEA 992 EUR
FLUO022876-1G
2-Phenyl-4'-(trifluormethyl)acetophenon
2-Phenyl-4'-(trifluormethyl)acetophenon
Formel: C₁₅H₁₁F₃O
MDL: MFCD02260678
CAS-Nummer: 61062-55-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR