4'-Chlor-3'-fluor-2-phenylacetophenon
Lieferant: FLUOROCHEM
022883-1G
FLUO022883-1GEA
1200
EUR
FLUO022883-1G
4'-Chlor-3'-fluor-2-phenylacetophenon
4'-Chlor-3'-fluor-2-phenylacetophenon
Formel:
C₁₄H₁₀ClFO |
MDL:
MFCD06201639 CAS-Nummer: 845781-23-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...