4'-Chlor-3'-methyl-2-phenylacetophenon

Lieferant: FLUOROCHEM

022884-1G
FLUO022884-1GEA 1200 EUR
FLUO022884-1G
4'-Chlor-3'-methyl-2-phenylacetophenon
4'-Chlor-3'-methyl-2-phenylacetophenon
Formel: C₁₄H₁₀ClFO
MDL: MFCD00196129
CAS-Nummer: 201990-40-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR