3',4'-Dichlor-2-phenylacetophenon

Lieferant: FLUOROCHEM

022886-1G
FLUO022886-1GEA 927 EUR
FLUO022886-1G
3',4'-Dichlor-2-phenylacetophenon
3',4'-Dichlor-2-phenylacetophenon
Formel: C₁₄H₁₀Cl₂O
MDL: MFCD06201641
CAS-Nummer: 93534-22-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR