3',5'-Dichlor-2-phenylacetophenon

Lieferant: FLUOROCHEM

022887-1G
FLUO022887-1GEA 1200 EUR
FLUO022887-1G
3',5'-Dichlor-2-phenylacetophenon
3',5'-Dichlor-2-phenylacetophenon
Formel: C₁₄H₁₀Cl₂O
MDL: MFCD06201642
CAS-Nummer: 845781-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR