3',4'-Dimethoxy-2-phenylacetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(3,4-Dimethoxyphenyl)-2-phenylethanone

022892-1G
FLUO022892-1GEA 0 EUR
FLUO022892-1G
3',4'-Dimethoxy-2-phenylacetophenon
3',4'-Dimethoxy-2-phenylacetophenon
Formel: C₁₆H₁₆O₃
Molecular Weight: 256,3000 g/mol
MDL: MFCD00219086
CAS-Nummer: 3141-93-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR