Desoxybenzoin
Lieferant: FLUOROCHEM
Synonyme:
2-Phenylacetophenone, 1,2-Diphenylethanone
022896-1G
FLUO022896-1GEA
56.7
EUR
FLUO022896-1G
Desoxybenzoin
Desoxybenzoin
Formel:
C₁₄H₁₂O Molecular Weight: 196,25 g/mol |
MDL:
MFCD00003081 CAS-Nummer: 451-40-1 EINECS: 207-193-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...