ω-(4-Fluorphenyl)acetophenon
Lieferant: FLUOROCHEM
022897-1G
FLUO022897-1GEA
461
EUR
FLUO022897-1G
ω-(4-Fluorphenyl)acetophenon
ω-(4-Fluorphenyl)acetophenon
Formel:
C₁₄H₁₁FO |
MDL:
MFCD02260684 CAS-Nummer: 347-91-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...