Benzyl-3-fluorphenylketon
Lieferant: FLUOROCHEM
Synonyme:
1-(3-Fluorophenyl)-2-phenylethanone
022909-1G
FLUO022909-1GEA
575
EUR
FLUO022909-1G
Benzyl-3-fluorphenylketon
Benzyl-3-fluorphenylketon
Formel:
C₁₄H₁₁FO Molecular Weight: 214,235 g/mol |
MDL:
MFCD02260663 CAS-Nummer: 40281-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...