Benzyl-4-fluorphenylketon
Lieferant: FLUOROCHEM
Synonyme:
1-(4-Fluorophenyl)-2-phenylethanone
022910-1G
FLUO022910-1GEA
38.6
EUR
FLUO022910-1G
Benzyl-4-fluorphenylketon
Benzyl-4-fluorphenylketon
Formel:
C₁₄H₁₁FO Molecular Weight: 214,2350 g/mol |
MDL:
MFCD00017960 CAS-Nummer: 347-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...