Ethyl(3-chlorbenzoyl)formiat
Lieferant: FLUOROCHEM
Synonyme:
Ethyl 2-(3-chlorophenyl)-2-oxoacetate
023000-1G
FLUO023000-1GEA
656
EUR
FLUO023000-1G
Ethyl(3-chlorbenzoyl)formiat
Ethyl(3-chlorbenzoyl)formiat
Formel:
C₁₀H₉ClO₃ Molecular Weight: 212,630 g/mol |
MDL:
MFCD01319616 CAS-Nummer: 62123-73-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...