Ethyl(3-fluor-4-methoxybenzoyl)formiat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-(3-fluoro-4-methoxyphenyl)-2-oxoacetate

023001-1G
FLUO023001-1GEA 336 EUR
FLUO023001-1G
Ethyl(3-fluor-4-methoxybenzoyl)formiat
Ethyl(3-fluor-4-methoxybenzoyl)formiat
Formel: C₁₁H₁₁FO₄
Molecular Weight: 226,201 g/mol
MDL: MFCD06201699
CAS-Nummer: 345-72-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR