Ethyl(3,4-dichlorbenzoyl)formiat
Lieferant: FLUOROCHEM
023002.1G
023002-25G
FLUO023002.1GEA
258
EUR
FLUO023002.1G
FLUO023002-25G
Ethyl(3,4-dichlorbenzoyl)formiat
Ethyl(3,4-dichlorbenzoyl)formiat
Formel:
C₁₀H₈Cl₂O₃ |
MDL:
MFCD01934864 CAS-Nummer: 34966-52-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...