Ethyl(4-methoxybenzoyl)formiat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 4-methoxybenzoylformate

023003-1G
FLUO023003-1GEA 84.9 EUR
FLUO023003-1G
Ethyl(4-methoxybenzoyl)formiat
Ethyl(4-methoxybenzoyl)formiat
Formel: C₁₀H₉ClO₃
Molecular Weight: 208.2106 g/mol
MDL: MFCD00963513
CAS-Nummer: 40140-16-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR