Ethyl(3,5-dichlorbenzoyl)formiat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-(3,5-dichlorophenyl)-2-oxoacetate

023004-1G
FLUO023004-1GEA 345 EUR
FLUO023004-1G
Ethyl(3,5-dichlorbenzoyl)formiat
Ethyl(3,5-dichlorbenzoyl)formiat
Formel: C₁₀H₈Cl₂O₃
Molecular Weight: 247,075 g/mol
MDL: MFCD06201700
CAS-Nummer: 845790-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR