Ethyl(3-methoxybenzoyl)formiat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-(3-methoxyphenyl)-2-oxoacetate

023005-1G
FLUO023005-1GEA 0 EUR
FLUO023005-1G
Ethyl(3-methoxybenzoyl)formiat
Ethyl(3-methoxybenzoyl)formiat
Formel: C₁₀H₉ClO₃
CAS-Nummer: 86358-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR