Ethyl-3,4-ethylendioxybenzoylformiat
Lieferant: FLUOROCHEM
Synonyme:
Ethyl 2-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)-2-oxoacetate
023006-1G
FLUO023006-1GEA
462
EUR
FLUO023006-1G
Ethyl-3,4-ethylendioxybenzoylformiat
Ethyl-3,4-ethylendioxybenzoylformiat
Formel:
C₁₂H₁₂O₅ Molecular Weight: 236,221 g/mol |
MDL:
MFCD06201701 CAS-Nummer: 131030-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...