Ethyl(4-phenoxybenzoyl)formiat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-oxo-2-(4-phenoxyphenyl)acetate

023007-1G
FLUO023007-1GEA 285 EUR
FLUO023007-1G
Ethyl(4-phenoxybenzoyl)formiat
Ethyl(4-phenoxybenzoyl)formiat
Formel: C₁₆H₁₄O₄
CAS-Nummer: 62936-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR