Ethyl-3,4-methylendioxybenzoylformiat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-(benzo[d][1,3]dioxol-5-yl)-2-oxoacetate

023009-1G
FLUO023009-1GEA 0 EUR
FLUO023009-1G
Ethyl-3,4-methylendioxybenzoylformiat
Ethyl-3,4-methylendioxybenzoylformiat
Formel: C₁₁H₁₀O₅
Molecular Weight: 222,194 g/mol
MDL: MFCD06201704
CAS-Nummer: 86358-30-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR