Ethyl(3,5-difluorbenzoyl)formiat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-(3,5-difluorophenyl)-2-oxoacetate

023011-1G
FLUO023011-1GEA 0 EUR
FLUO023011-1G
Ethyl(3,5-difluorbenzoyl)formiat
Ethyl(3,5-difluorbenzoyl)formiat
Formel: C₁₀H₈F₂O₃
Molecular Weight: 214,17 g/mol
MDL: MFCD01319620
CAS-Nummer: 208259-57-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR