Ethyl(4-butylbenzoyl)formiat
Lieferant: FLUOROCHEM
023012-1G
FLUO023012-1GEA
0
EUR
FLUO023012-1G
Ethyl(4-butylbenzoyl)formiat
Ethyl(4-butylbenzoyl)formiat
Formel:
C₁₄H₁₈O₃ Molecular Weight: 234,291 g/mol |
CAS-Nummer:
80120-35-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...