Ethyl(3,4,5-trifluorbenzoyl)formiat

Lieferant: FLUOROCHEM

023013-1G
FLUO023013-1GEA 647 EUR
FLUO023013-1G
Ethyl(3,4,5-trifluorbenzoyl)formiat
Ethyl(3,4,5-trifluorbenzoyl)formiat
Formel: C₁₀H₇F₃O₃
CAS-Nummer: 732251-58-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR