Ethyl(3,5-dimethylbenzoyl)formiat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-(3,5-dimethylphenyl)-2-oxoacetate

023014-1G
FLUO023014-1GEA 434 EUR
FLUO023014-1G
Ethyl(3,5-dimethylbenzoyl)formiat
Ethyl(3,5-dimethylbenzoyl)formiat
Formel: C₁₂H₁₄O₅
Molecular Weight: 206,238 g/mol
MDL: MFCD01311166
CAS-Nummer: 100117-62-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR