Ethyl(4-tert-butylbenzoyl)formiat

Lieferant: FLUOROCHEM

023015-1G
FLUO023015-1GEA 0 EUR
FLUO023015-1G
Ethyl(4-tert-butylbenzoyl)formiat
Ethyl(4-tert-butylbenzoyl)formiat
Formel: C₁₄H₁₈O₃
CAS-Nummer: 80120-36-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR