Ethyl(3,4-dimethylbenzoyl)formiat

Lieferant: FLUOROCHEM

023016-1G
FLUO023016-1GEA 0 EUR
FLUO023016-1G
Ethyl(3,4-dimethylbenzoyl)formiat
Ethyl(3,4-dimethylbenzoyl)formiat
Formel: C₁₂H₁₄O₅
CAS-Nummer: 80120-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR