Ethyl(3-chlor-4-fluorbenzoyl)formiat

Lieferant: FLUOROCHEM

023017-1G
FLUO023017-1GEA 115 EUR
FLUO023017-1G
Ethyl(3-chlor-4-fluorbenzoyl)formiat
Ethyl(3-chlor-4-fluorbenzoyl)formiat
Formel: C₁₀H₈ClFO₃
CAS-Nummer: 845790-53-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR