Ethyl(3-isopropoxybenzoyl)formiat

Lieferant: FLUOROCHEM

023018-1G
FLUO023018-1GEA 515 EUR
FLUO023018-1G
Ethyl(3-isopropoxybenzoyl)formiat
Ethyl(3-isopropoxybenzoyl)formiat
Formel: C₁₀H₉ClO₃
CAS-Nummer: 845790-54-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR