Ethyl(4-propylbenzoyl)formiat

Lieferant: FLUOROCHEM

023020-1G
FLUO023020-1GEA 554 EUR
FLUO023020-1G
Ethyl(4-propylbenzoyl)formiat
Ethyl(4-propylbenzoyl)formiat
Formel: C₁₃H₁₆O₃
CAS-Nummer: 845790-55-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR